missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Multi-Compendial, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Formula Weight | 75.07 |
Grade | Multi-Compendial/USP |
Quantity | 2 kg |
Specific Gravity | 1L = 1.16kg |
Chemical Name or Material | Glycine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'