missing translation for 'onlineSavingsMsg'
Learn More

Glycine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7081954
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
6000-43-7
C2H6ClNO2
111.53
MFCD00012872
IVLXQGJVBGMLRR-UHFFFAOYSA-N
glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride
22316
2-aminoacetic acid;hydrochloride
[H+].[Cl-].NCC(O)=O
This item is not returnable. View return policy
6000-43-7
C2H6ClNO2
111.53
MFCD00012872
IVLXQGJVBGMLRR-UHFFFAOYSA-N
glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride
22316
2-aminoacetic acid;hydrochloride
[H+].[Cl-].NCC(O)=O

Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Soluble in water (>100 g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY
Melting Point 175°C to 178°C
Beilstein 3909617
Merck Index 14,4491
Solubility Information Soluble in water (>100g/L).
Formula Weight 111.53
Percent Purity 98%
Quantity 100 g
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Glycine hydrochloride
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'