missing translation for 'onlineSavingsMsg'
Learn More

Glycine anhydride, 98%, Thermo Scientific Chemicals

Catalog Number 10513314
859.50 GBP valid until 2024-03-30
Use promo code "21648" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
106-57-0
C4H6N2O2
114.1
MFCD00006009
BXRNXXXXHLBUKK-UHFFFAOYSA-N
2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
7817
CHEBI:16535
piperazine-2,5-dione
C1C(=O)NCC(=O)N1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
106-57-0
C4H6N2O2
114.1
MFCD00006009
BXRNXXXXHLBUKK-UHFFFAOYSA-N
2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
7817
CHEBI:16535
piperazine-2,5-dione
C1C(=O)NCC(=O)N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color White to Yellow
Loss on Drying 0.5% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex N)
Packaging Glass bottle
Quantity 1 kg
Beilstein 24, 264
Merck Index 15, 7578
Solubility Information Solubility in water: 142g/L water (20 c). Other solubilities: soluble in hcl, slightly soluble in ethanol
Formula Weight 114.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Glycine anhydride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'