missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 99+%, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog Number 15474789
Click to view available options
Quantity:
100 g
500 g
1 kg
56-40-6
C2H5NO2
75.07
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
NCC(O)=O
56-40-6
C2H5NO2
75.07
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
NCC(O)=O

This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
Soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature 2 - 8°C
TRUSTED_SUSTAINABILITY
Melting Point ∽245°C (decomposition)
pH 5.97
Color White
Linear Formula H2NCH2CO2H
Beilstein 635782
Merck Index 14,4491
Solubility Information Soluble in water (100mg/ml),and acetone (slightly soluble). Insoluble in alcohol,and ether.
Formula Weight 75.07
Percent Purity ≥99.5%
Grade Cell Culture Reagent
Density 1.595
Quantity 1 kg
Odor Odorless
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'