Learn More
Merck Millipore Glycine
Guaranteed Reagent (GR) for analysis
Brand: Merck Millipore 1042010250
Code : NEW
Additional Details : CAS Number : 56-40-6 Weight : 0.25000kg
Quantity | 250 g |
---|
Description
- Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
- Analysis Note:
- Assay (perchloric acid titration): ≥ 99.7 %
- Identity (IR-spectrum): passes test
- pH (50 g/L CO2-free water): 5.9 - 6.3
- Chloride (Cl): ≤ 0.003 %
- Sulfate (SO4): ≤ 0.0025 %
- Heavy metals (as Pb): ≤ 0.001 %
- Cu (Copper): ≤ 0.0001 %
- Fe (Iron): ≤ 0.0001 %
- Pb (Lead): ≤ 0.0001 %
- NH4 (Ammonium): ≤ 0.02 %
- Foreign amino acids: ≤ 0.1 %
- Other ninhydrine positive substances: ≤ 0.1 %
- In water insoluble matter: ≤ 0.003 %
- Sulfated ash (600°C): ≤ 0.05 %
Chemical Identifiers
≥99.7% (perchloric acid titration) | |
7930 mg/kg LD50, oral (Rat) | |
1 S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5) | |
NH2CH2COOH | |
75.07 | |
Solid | |
250 g/L |
56-40-6 | |
1.161 g/cm3 (at 20°C) | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
MFCD00008131 | |
5.9-6.4 (20°C, 50 g/L in H2O) | |
NCC(O)=O | |
Aminoacetic acid, Aminoethanoic acid, Gly, Aminoacetic acid, Aminoethanoic acid, Glycocoll |
Specifications
56-40-6 | |
C2H5NO2 | |
MFCD00008131 | |
1 S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5) | |
NCC(O)=O | |
75.07 | |
250 g/L | |
5.9-6.4 (20°C, 50 g/L in H2O) | |
7930 mg/kg LD50, oral (Rat) |
≥99.7% (perchloric acid titration) | |
NH2CH2COOH | |
Aminoacetic acid, Aminoethanoic acid, Gly, Aminoacetic acid, Aminoethanoic acid, Glycocoll | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
Solid | |
1.161 g/cm3 (at 20°C) | |
250 g |