missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycinamide hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.
Solubility
Soluble in water (1100g/L).
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.
Solubility
Soluble in water (1100g/L).
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | ∼204°C (decomposition) |
Quantity | 5 g |
Beilstein | 3554199 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water (1100g/L). |
Formula Weight | 110.54 |
Percent Purity | 98% |
Chemical Name or Material | Glycinamide hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'