missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl 2-methoxyphenyl ether, 98%, Thermo Scientific™

Catalog Number 15438348
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2210-74-4
C10H12O3
180.203
MFCD00190158
RJNVSQLNEALZLC-UHFFFAOYSA-N
1-2-methoxyphenoxy-2,3-epoxypropane, guaiacol glycidyl ether, 2-2-methoxyphenoxy methyl oxirane, guajacol-glycidaether, 1,2-epoxy-3-o-methoxyphenoxy propane, glycidyl 2-methoxyphenyl ether, 1,2-epoxy-3-2-methoxyphenoxy propane, oxirane, 2-methoxyphenoxy methyl, guajacol-glycidaether german
98357
2-[(2-methoxyphenoxy)methyl]oxirane
COC1=CC=CC=C1OCC2CO2
This item is not returnable. View return policy
2210-74-4
C10H12O3
180.203
MFCD00190158
RJNVSQLNEALZLC-UHFFFAOYSA-N
1-2-methoxyphenoxy-2,3-epoxypropane, guaiacol glycidyl ether, 2-2-methoxyphenoxy methyl oxirane, guajacol-glycidaether, 1,2-epoxy-3-o-methoxyphenoxy propane, glycidyl 2-methoxyphenyl ether, 1,2-epoxy-3-2-methoxyphenoxy propane, oxirane, 2-methoxyphenoxy methyl, guajacol-glycidaether german
98357
2-[(2-methoxyphenoxy)methyl]oxirane
COC1=CC=CC=C1OCC2CO2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 41°C to 48°C
Color White
Boiling Point 152°C to 158°C
Quantity 5 g
Formula Weight 180.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Glycidyl 2-methoxyphenyl ether
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'