missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycidyl 2,2,3,3-tetrafluoropropyl ether, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Glycidyl 2,2,3,3-tetrafluoropropyl ether is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Glycidyl 2,2,3,3-tetrafluoropropyl ether is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.327 |
Boiling Point | 50°C (4 mmHg) |
Flash Point | 79°C (174°F) |
Refractive Index | 1.366 |
Quantity | 25 g |
Formula Weight | 188.12 |
Percent Purity | 97% |
Chemical Name or Material | Glycidyl 2,2,3,3-tetrafluoropropyl ether |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'