missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Solution, 85%, Honeywell™ Riedel-de-Haën™

Product Code. 15625700
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15625700

Brand: Honeywell-Riedel-de Haen 497831L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tested according to European Pharmacopoeia, 85%

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 56-81-5
Color Colorless
Density 1.252g/mL at 25°C
pH Neutral
Boiling Point 290°C (1013 hPa)
Flash Point 160°C (320°F)
Odor Odorless
Packaging Glass bottle
Molecular Formula C3H8O3
Linear Formula HOCH2CH(OH)CH2OH · aq
MDL Number MFCD00004722
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
Solubility Information Completely miscible in water
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES OCC(O)CO
IUPAC Name propane-1,2,3-triol
Molecular Weight (g/mol) 92.09
PubChem CID 753
ChEBI CHEBI:17754
Formula Weight 92.09g/mol (anhydrous basis)
Physical Form Liquid
Chemical Name or Material Glycerol solution, Tested according to Ph.Eur.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.