Learn More
Glycerol monostearate, purified, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Glycerol monostearate is used as an emulsifier, resin lubricant, opacifier, emollient, bodying agent in a variety of cosmetic formulations for skincare and haircare. It is also used as a thickening, anti-caking and preservative agent. It is also useful for preventing ice creams from drying out or being too sweet. It is further used as a foaming agent for the foam-mat drying of papaya. It is also used as an anti-staling agent in bread.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 250 g |
Merck Index | 14,4489 |
Solubility Information | Soluble in hot organic solvents;Soluble in hot water; Slightly soluble in ethanol; Insoluble in aliphatic solvents. |
Formula Weight | 358.56 |
Physical Form | Powder |
Chemical Name or Material | Glycerol monostearate, Purified |
RUO – Research Use Only