missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™

Catalog Number 10214580
1364.25 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO

C3H8O3, CAS Number-56-81-5, glycerol, propanetriol, glycyl alcohol, glycerin, 1,2,3-trihydroxypropane, glycerine, 1,2,3-propanetriol, trihydroxypropane, osmoglyn, glyceritol, 1L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722

TRUSTED_SUSTAINABILITY
Melting Point 18°C
pH 5
Boiling Point 290°C
Packaging Plastic container
Quantity 25 L
Vapor Density 3.17
Formula Weight 92.09g/mol
Vapor Pressure 0.003mbar 50
Physical Form Very Viscous Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'