missing translation for 'onlineSavingsMsg'
Learn More

Glycerol-1-allylether, 98%, ACROS Organics™

Brand:  Acros Organics 240671000

Code : 29

Additional Details : CAS Number : 123-34-2 Weight : 0.15700kg


Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10730765

  • £19.70 / 100mL


Estimated Shipment
19-03-2018
Add to basket


Specifications

Chemical Identifiers

Glycerol-1-allylether
98%
CH2=CHCH2OCH(OH)CH2OH
MFCD00004721
98%
C6H12O3
132.16
3-(2-Propenyloxy)-1, 2-propanediol

Specifications

Glycerol-1-allylether
123-34-2
98%
1.0700g/mL
CH2=CHCH2OCH(OH)CH2OH
MFCD00004721
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, acetone and toluene
Glass bottle
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, acetone and toluene
Clear colorless to light yellow
132°C
100mL
Viscous Liquid
98%
C6H12O3
132.16
3-(2-Propenyloxy)-1, 2-propanediol
Authentic
1.4610 to 1.4630
1.07
245.0°C
Videos
SDS