missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerin, 100% Vegetable Based, Multi-Compendial, U.S.P., J.T. Baker™
266.95 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
Specifications
Flash Point | 199.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 500 mL |
Specific Gravity | 1L = 1.26kg |
Formula Weight | 92.1 |
Percent Purity | 100% Vegetable Based |
Grade | Multi-Compendial/USP |
Chemical Name or Material | Glycerin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'