missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, 100% Vegetable Based, Multi-Compendial, U.S.P., J.T. Baker™

Catalog Number 15517984
266.95 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
This item is not returnable. View return policy
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO
This item is not returnable. View return policy
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO
Flash Point 199.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.26kg
Formula Weight 92.1
Percent Purity 100% Vegetable Based
Grade Multi-Compendial/USP
Chemical Name or Material Glycerin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'