Learn More
Glutaric dialdehyde, 50 wt.% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 410960100
Code : 29
Additional Details : CAS Number : 111-30-8 Weight : 10.00000kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glutaric dialdehyde | |
111-30-8,7732-18-5 | |
1.1300g/mL | |
101.5°C (740.0 mmHg) | |
C5H8O2 | |
MFCD00007025 | |
01, 776 | |
1.13 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
CHEBI:64276 | |
Solution |
50 wt% Solution in Water | |
-33.0°C | |
3.2 to 4.2 | |
Plastic drum | |
OHC(CH2)3CHO | |
10 kg | |
01,411 | |
15, 4508 | |
Solubility in water: soluble | |
O=CCCCC=O | |
100.12 | |
20 mPa.s (50°C) | |
100.12 |
RUO – Research Use Only