missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose-1-Phosphoric Acid, Dipotassium Salt Dihydrate, Fisher Chemical
Click to view available options
Quantity:
2 g
Packaging:
Vial
Specifications
CAS | 5996-14-5 |
Color | White |
Physical Form | Solid |
Quantity | 2 g |
Molecular Formula | C6H17K2O11P |
MDL Number | 149435 |
Synonym | alpha-d-glucopyranose, 1-dihydrogen phosphate , dipotassium salt, dihydrate, alpha-d-glucopyranose 1-phosphate dipotassium salt hydrate, alpha-d-glucose 1-phosphate dipotassium salt dihydrate, alpha-glucose-1-phosphate dipotassium dihydrate, alpha-glucose-1-phosphate dipotassium salt dihydrate mi, a-d-glucose-1-phosphate dipotassium salt hydrate, alpha-d-glucose-1-phosphate dipotassium salt hydrate, dipotassium dihydrate ?-d-glucopyranosyl phosphate, dipotassium 2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl phosphate dihydrate, potassium 2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl phosphate dihydrate |
InChI Key | RZULLROGARPECB-QMKHLHGBSA-N |
SMILES | C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.O.O.[K].[K] |
IUPAC Name | potassium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate;dihydrate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'