missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucosamine (N-acetyl)-6-Sulfatase/GNS Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Glucosamine (N-acetyl)-6-Sulfatase/GNS |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Glucosamine (N-acetyl)-6-Sulfatase/GNS Val37-Leu552 Accession # P15586 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
Glucosamine (N-acetyl)-6-Sulfatase/GNS Polyclonal specifically detects Glucosamine (N-acetyl)-6-Sulfatase/GNS in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Glucosamine (N-acetyl)-6-Sulfatase/GNS | |
Goat | |
Polyclonal | |
IgG | |
EC 3.1.6, EC 3.1.6.14, G6Sglucosamine-6-sulfatase, glucosamine (N-acetyl)-6-sulfatase, Glucosamine-6-sulfatase, Glucosamine6Sulfatase, GNS, MGC21274, N-acetylglucosamine-6-sulfatase | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Immunoprecipitation | |
RUO |
Mouse myeloma cell line NS0-derived recombinant human Glucosamine (N-acetyl)-6-Sulfatase/GNS Val37-Leu552 Accession # P15586 | |
Primary | |
Human | |
Unconjugated | |
2799 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only