missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucokinase/GCK Antibody (223), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89787F
This item is not returnable.
View return policy
Description
Glucokinase/GCK Monoclonal specifically detects Glucokinase/GCK in Human samples. It is validated for ELISA.
Specifications
| Glucokinase/GCK | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 223 | |
| ELISA | |
| EC 2.7.1, EC 2.7.1.2, FGQTL3, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Glucokinase/GCK (NP_000153.1; Leu2-Gln465). | |
| 0.1 mL | |
| Lipid and Metabolism, Protein Kinase | |
| 2645 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction