missing translation for 'onlineSavingsMsg'
Learn More

Glucagon Mouse anti-Human, Mouse, Rat, Biotin, Clone: ICACLS, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9743-80

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15567366

  • £104.00 / 25µg

Estimated Shipment
27-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Glucagon is a 29-residue polypeptide hormone (MW 3482), produced in the pancreas. A related hormone, enteroglucagon (or oxyntomodulin), which is produced in the mucosa of the small and large intestine, consists of the 29 amino acid sequence of pancreatic glucagon extended by 8 additional residues at the C-terminus. The biological activities of pancreatic glucagon include glycogenolysis, lipolysis, gluconeogenesis, and ketogensis, which are antagonistic effects to those of insulin action, thus leading to increased blood glucose levels. Immunocytochemical studies have revealed the presence of pancreatic glucagon inside the A or alpha cells, which constitute 15-20% of the islet cell population. These cells are located preferentially at the periphery of the human pancreatic islets. Pathological manifestations of the glucagon-type peptide residue almost exclusively with the exsistence of tumors or glucagonomas, as no states of glucagon-cell deficiency or hyperplasia have been identified. Glucagon-specific antibodies would prove useful as a cell and tumor markers applying immunohistochemical techniques, and as an analytical tool in qualification of the hormone.
Specifications

Specifications

Glucagon
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06883, P01275, P55095
GCG, GLP1, GLP2, GRPP
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
ICACLS
Biotin
GCG
Liquid
GCG
IgG1, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14526, 24952, 2641
Videos
Documents

For Research Use Only.