missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Glove Bag Clips

Catalog Number 15431847
Click to view available options
Quantity:
36 Pk.
6 Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

For convenient hanging of glove storage bags.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • For convenient hanging of glove storage bag
  • Available in packs of 6 or 36
TRUSTED_SUSTAINABILITY
Content And Storage Ambient temperatures
For Use With (Equipment) Glove Storage Bags
Quantity 6 Pk.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'