missing translation for 'onlineSavingsMsg'
Learn More

Glimepiride, Tocris Bioscience™

Kir6 (KATP) channel blocker

Brand:  Tocris (Bio-Techne) 239610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441134

  • £67.62 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent Kir6 (KATP) channel blocker and anti-diabetic agent. Inhibits pinacidil-activated cardiac Kir6 channels with an IC50 of 6.8 nM.
Specifications

Chemical Identifiers

93479-97-1
490.619
glimepiride, amaryl, glimepiridum, glimepirida, amarel, glimepirid, glimepride, endial, roname, glimepiridum latin
4-ethyl-3-methyl-N-[2-[4-[(4-methylcyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-5-oxo-2H-pyrrole-1-carboxamide
C24H34N4O5S
WIGIZIANZCJQQY-UHFFFAOYSA-N
3476
CCC1=C(CN(C1=O)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCC(CC3)C)C

Specifications

10mg
93479-97-1
glimepiride, amaryl, glimepiridum, glimepirida, amarel, glimepirid, glimepride, endial, roname, glimepiridum latin
CCC1=C(CN(C1=O)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCC(CC3)C)C
490.619
490.62
Glimepiride
C24H34N4O5S
WIGIZIANZCJQQY-UHFFFAOYSA-N
4-ethyl-3-methyl-N-[2-[4-[(4-methylcyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-5-oxo-2H-pyrrole-1-carboxamide
3476
>98%
Videos
Documents