missing translation for 'onlineSavingsMsg'
Learn More

GFAP Monoclonal Antibody (GA5), eFluor™ 615, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 42-9892-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15301130

  • £155.00 / 25µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This GA5 monoclonal antibody reacts with human, mouse, rat, chicken, rabbit, and pig glial fibrillary acidic protein (GFAP). This 49-kDa type III intermediate filament protein is expressed in neural tissues and distinguishes astrocytes from other glial cells during central nervous system development. Three alternative splice variants of GFAP exist; however, alpha-GFAP is the predomit form expressed in astrocytes. GFAP can co-assemble with vimentin and nestin in astrocytes, but such associations are not required for assembly. Like other intermediate filaments, GFAP assembly is dependent on phosphorylation and dephosphorylation of the N-terminal domain. Studies have demonstrated that mutations in the GFAP gene lead to Alexander disease. Moreover, GFAP has also been shown to be overexpressed in certain glial-derived tumors. Applications Reported: This GA5 antibody has been reported for use in immunocytochemical (ICC) and immunohistochemical staining of frozen tissue sections (IHC-F). Applications Tested: This GA5 antibody has been tested by immunocytochemistry on fixed and permeabilized C6 cells at less than or equal to 10 μg/mL. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor™ 615 antibody conjugate, we recommend a filter that will capture the 615 emission wavelength (for example, Excitation 560/55, 585LP, Emission 645/75).

GFAP Monoclonal antibody specifically detects GFAP in Chicken, Human, Mouse, Porcine, Rabbit, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

GFAP
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P03995, P14136, P47819
GFAP
Affinity chromatography
RUO
100352532, 14580, 24387, 2670, 396562, 419969
4° C, store in dark, DO NOT FREEZE!
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen)
GA5
eFluor 615
GFAP
6330404F12Rik; 65 kDa glutamic acid decarboxylase; AI836096; ALXDRD; Astrocyte or Intermediate Filament Protein; cb345; class III intermediate filament protein; etID36982.3; FLJ45472; GAD(65); Gad2; Gad-2; GAD65; GAD-65; GFAP; GFAP epsilon; gfap protein; gfapl; glial fibrillary acidic protein; Glial Fibrillary Acidic Protein (GFAP); glial fibrillary acidic protein alpha; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; glutamic acid decarboxylase 2; I79_011607; intermediate filament; intermediate filament protein; intermediate filament protein; GFAP; Unknown (protein for MGC:139638); wu:fb34h11; wu:fk42c12; xx:af506734; zgc:110485; zGFAP; intermediate filament protein; zrf-1 antigen
Mouse
25 μg
Primary
Chicken, Human, Mouse, Porcine, Rabbit, Rat
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.