missing translation for 'onlineSavingsMsg'
Learn More

Glial Fibrillary Acid Protein (GFAP) Mouse, eFluor 570, Clone: GA5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 41-9892-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15548986

  • £172.54 / 100µg


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

This GA5 monoclonal antibody reacts with human, mouse, rat, chicken, rabbit, and pig glial fibrillary acidic protein (GFAP). This 49kDa type III intermediate filament protein is expressed in neural tissues and distinguishes astrocytes from other glial cells during central nervous system development. Three alternative splice variants of GFAP exist; however, α-GFAP is the predominant form expressed in astrocytes. GFAP can co-assemble with vimentin and nestin in astrocytes, but such associations are not required for assembly. Like other intermediate filaments, GFAP assembly is dependent on phosphorylation and dephosphorylation of the N-terminal domain. Studies have demonstrated that mutations in the GFAP gene lead to Alexander disease. Moreover, GFAP has also been shown to be overexpressed in certain glial-derived tumors.
Specifications

Specifications

Glial Fibrillary Acid Protein (GFAP)
GA5
eFluor 570
Conjugated
IgG1
RUO
Primary
Rat
Microscopy
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos