Learn More
GLB1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560221
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GRYWTKQGPQ QTLYVPRFLL FPRGALNKIT LLELEDVPLQ PQVQFLDKPI LNSTSTLHRT HINSLSADTL SASEPMELSG H Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 67%.
GLB1L is a probable glycosyl hydrolase (Potential).Specifications
GLB1L | |
Polyclonal | |
Unconjugated | |
GLB1L | |
4833408P15Rik; Beta-galactosidase-1-like protein; galactosidase beta 1 like; galactosidase beta 1-like; galactosidase, beta 1-like; Glb1l; testicular tissue protein Li 74; UNQ229/PRO262 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79411 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWU2 | |
GLB1L | |
Recombinant protein corresponding to Human GLB1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only