missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Glassy Carbon Tapered Crucible for Crystal Growth

Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.

Brand:  Alfa Aesar™ 040955.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15493745

  • £454.95 / Each


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Specifications

37mL
Top: 51mm
30mm
Glassy Carbon
Tapered
For Crystal Growth
Glassy Carbon Crucible
Videos
SDS