missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Glassy Carbon E-Beam Evaporating Crucible

Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.

Brand:  Alfa Aesar™ 040964.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15483755

  • £498.20 / Each


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Specifications

16mL
37mm
E-Beam Evaporating
Glassy Carbon Crucible
Angle: 15 deg.
47mm
18mm
Glassy Carbon
Videos
SDS