missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Glassy Carbon Cylindrical Crucible

Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.

Brand:  Alfa Aesar™ 039013.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15426285

  • £651.58 / Each


Estimated Shipment date
from Supplier 21-06-2018
Add to basket

Specifications

Specifications

140mL
47mm
Glassy Carbon Crucible
Cylindrical
104mm
Glassy Carbon
Videos
SDS