missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Glass Wide Neck Flask

Brand:  Duran™ 217313602

Code : A0

Additional Details : Weight : 1.30000kg

 View more versions of this product

Product Code. 11790764

  • £84.20 / Pack of 10


Estimated Shipment
20-07-2018
Add to basket

Specifications

Specifications

Yes
DIN ISO 24450, USP Standard
250mL
Ideal as Heating Application
Wide
Flat
85mm
Glass
138mm
Videos