missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Glass Wide Neck Flask

Brand:  Duran™ 217312409

Code : A0

Additional Details : Weight : 0.70000kg

 View more versions of this product

Product Code. 11780764

  • £78.40 / Pack of 10


Estimated Shipment
02-03-2018
Add to basket

Specifications

Specifications

Yes
DIN ISO 24450, USP Standard
100mL
Ideal as Heating Application
Wide
Flat
64mm
Glass
105mm
Videos