missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Glass Trapezoidal Volumetric Flask with Polyethylene Stopper

Brand:  Hirschmann Instruments™ 2960170

Code : A0

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11796752

  • £36.10 / Pack of 2


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

DIN EN ISO 1042
Polyethylene
Borosilicate GlassþDURAN™
110mm
10/19 NS Standard Ground
Class A
25mL
Yes
±0.04mL
±0.04mL
Videos