missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Glass Trapezoidal Volumetric Flask with Polyethylene Stopper

Manufactured using class A, DURAN™ glass

Brand:  Hirschmann™ 2960152

Code : A0

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11766742

  • £40.45 / Pack of 2
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

  • Blue graduation
  • Certified conformity, with dated batch identification 
  • Trapezoidal
  • Standard ground and PE stopper
  • Calibrated for IN, DIN EN ISO 1042
Specifications

Specifications

DIN EN ISO 1042
5 mL
70 mm
7/16 NS Standard Ground
Yes
Polyethylene
Borosilicate Glass, Duran Glass
±0.025 mL
±0.025 mL
A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers