missing translation for 'onlineSavingsMsg'
Learn More

Buchi Thimble adapter 33mm x 94mm

Brand:  Buchi 037279

Thimble adapter 33mm x 94mm

Code : 62

Additional Details : Weight : 0.07000kg

Product Code. 12055472

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.