missing translation for 'onlineSavingsMsg'
Learn More

Buchi Thimble adapter 43mm x 123mm

Brand:  Buchi 037280

Thimble adapter 43mm x 123mm

Code : 62

Additional Details : Weight : 0.08000kg

Product Code. 12065472

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.