Learn More
Hirschmann Instruments™ Hollow Glass Stoppers
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
Brand:  Hirschmann Instruments™ 8900365
UNSPSC : 41120000
Code : A0
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 11716243
  • £33.05 / Pack of 10
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Description
  • Acc. to DIN 12 252
  • Available to fit a range of sizes
Description & Specifications
Specifications
Stopper
NS 12/21
Glass
1Each
For tight sealing of standard ground vessels
Alternative Products