Buchi™ Glass Soxhlet Extraction Chamber
Overview
Brand: Buchi™
Manufacturer Part Number: 018241
Soxhlet Extractor D30mm
UNSPSC: 41104812
Code: 62
Additional Details:
Additional Details: Weight: 0.21000kg
Product Code. 12830156
Quantity Price
1 £ 1468.65 / Each
Estimated Shipment
07-04-2017
Add to basket
Description and Specification
Specification
Material Glass
Diameter (Metric) Inner Chamber 30mm
Diameter (Metric) Inside 30mm
Type Soxhlet Extraction Chamber
For Use With For extraction