missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Disposable Micro Capillary Pipettes, End-to-End

Facilitates error-free capillary filling in the laboratory. Hirschmann Instruments™ Disposable Micro Capillary Pipettes, End-to-End are manufactured from durable DURAN™ glass.

Brand:  Hirschmann Instruments™ 9000201

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11767663

  • £86.00 / Pack of 1000


Estimated Shipment
22-05-2018
Add to basket

Description

Description

  • End-to-end, with minicaps
  • Conformity certified
  • Available either heparinized or non-heparinized
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications

Specifications

0.5%
Yes
Minicap Capillary Pipette
Glass
ISO 7550, CE, IVD 98/79 EC
30 to 32mm
1μL
Videos