missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Glass Serological Pipettes
Offering economical single-use convenience leading to greatly reduced possibility of cross-contamination. Poulten Graf™ Glass Serological Pipettes are disposable and have clear advantages over plastic counterparts in areas of chemical and temperature resistance, accuracy, transparency and clarity of markings.
£278.00 - £379.00
Specifications
Material | Glass |
---|---|
Graduated | Yes |
Quantity | 500/Pk. |
Type | Serological Pipette |
Disposable | Single-use |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11788245
|
Poulten Graf™
D803 |
2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12394438
|
Poulten Graf™
D807 |
5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Ideal for use with radioactive and other hazardous materials
- Environmentally friendly compared with plastic counterparts
- Black graduations, non-sterile
- Accurate to ± 2% of volume adjusted for blow out delivery
Specifications
Glass | |
500/Pk. | |
Single-use |
Yes | |
Serological Pipette |