Learn More
Poulten Graf™ Glass Serological Pipettes
Offering economical single-use convenience leading to greatly reduced possibility of cross-contamination. Poulten Graf™ Glass Serological Pipettes are disposable and have clear advantages over plastic counterparts in areas of chemical and temperature resistance, accuracy, transparency and clarity of markings.
Brand: Poulten Graf™ D803
Code : 02
Additional Details : Weight : 0.80000kg
Description
- Ideal for use with radioactive and other hazardous materials
- Environmentally friendly compared with plastic counterparts
- Black graduations, non-sterile
- Accurate to ± 2% of volume adjusted for blow out delivery
Specifications
Glass | |
0.02 mL | |
2 mL | |
Serological Pipette |
Yes | |
500/Pk. | |
270 mm | |
Single-use |