missing translation for 'onlineSavingsMsg'
Learn More

Buchi Tube with 3mL residual volume (graduated) for Crystal Analyst rack R-12 12 x 120mL

Brand:  Buchi 046070

Tube with 3mL residual volume (graduated) for Crystal Analyst rack R-12 12 x 120mL

Code : 62

Additional Details : Weight : 2.04000kg

Product Code. 12836723

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.