missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Glass Ribbed Funnels

Brand:  Duran™ 213524601

Code : A0

Additional Details : Weight : 1.01000kg

 View more versions of this product

Product Code. 11747981

  • £303.00 / Pack of 10


Estimated Shipment
08-06-2018
Add to basket

Specifications

Specifications

USP Standard
10mm
Ideal for filtering with round-paper
Glass
150 to 185mm
100mm
180mm
Videos