missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Glass Printed Scale Volumetric Flask

Brand:  Hirschmann Instruments™ 2950180

Code : A0

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11827112

  • £161.00 / Pack of 2


Estimated Shipment
18-07-2018
Add to basket

Specifications

Specifications

Class A
Borosilicate GlassþDURAN™
170mL
100mL
Yes
Videos