missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Glass Powder Funnels

Manufactured from Borosilicate glass. DURAN™ Glass Powder Funnels keep your work area free from spills.

Brand:  Duran™ 213544603

Code : A0

Additional Details : Weight : 0.82000kg

 View more versions of this product

Product Code. 11767981

  • £167.00 / Pack of 10


Estimated Shipment
16-02-2018
Add to basket

Description

Description

  • Duran™ powder funnel
  • Short, wide stem
  • Borosilicate glass
Specifications

Specifications

USP Standard
100mm
94mm
26mm
For decanting of powdered substances and granulated material
Glass
Videos