missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Glass Narrow Neck Flask

Brand:  Duran™ 217116406

Code : A0

Additional Details : Weight : 4.80000kg

 View more versions of this product

Product Code. 11780744

  • £341.00 / Pack of 10


Estimated Shipment
11-06-2018
Add to basket

Specifications

Specifications

Yes
ISO 1773, USP Standard
2000mL
Ideal as Heating Application
Narrow
Flat
166mm
Glass
250mm
Videos