Learn More
Kinematica™ Mixing Attachment for Microtron™ MB 550 Laboratory Mixer
For the mixing, dispersing, homogenizing, emulsifying and size reduction of cuttable materials.
Brand: Kinematica™ 36050010
Code : 04
Additional Details : Weight : 0.50000kg
Description
- For use with the MB 550 Mixers
- The two stainless steel ball bearings are sealed twice each
- Noiseless and sturdy coupling with dampers
- Blades, sealing and dampers can easily be exchanged
- Four high-grade blades designed for optimal mixing
- Generally suited for intensive and fast processing
Specifications
Mixing Head Attachment | |
Stainless Steel | |
Consists of gasket and blade head | |
1 Ea. |
250 mL | |
Borosilicate Glass | |
MB 550 Laboratory Mixer |