Learn More
DWK Life Sciences DURAN™ Funnel, with long stem, Bunsen Funnel
A funnel enables a clean decanting and filtering of liquids. In addition, the Bunsen funnel has a long handle.
Brand: DWK Life Sciences 213533803
Code : A0
Additional Details : Weight : 0.40000kg
Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- With long stem
- Bunsen funnel
- Fired-on, highly durable white print
Specifications
Non-DIN/ISO size | |
Boro 3.3 | |
Decanting and filtration of liquids of different densities; The spillage of substances is avoided; Filtering |
70 mm | |
Yes | |
10/Pk. |