missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Glass Funnels with Short Stem

Brand:  Duran™ 213516601

Code : A0

Additional Details : Weight : 0.87000kg

 View more versions of this product

Product Code. 10589671

  • £145.00 / Each


Estimated Shipment
31-01-2018
Add to basket

Description

Description

  • Manufactured from borosilicate glass
  • With short stem
Specifications

Specifications

ISO 4798, USP Standard
30mm
For decanting and filtration of substances
Glass
385 to 400mm
250mm
370mm
Videos