Learn More
Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
Brand:  Hirschmann Instruments™ 9600122
Code : NEW
Additional Details : Weight : 1.00000kg
Product Code. 10612242
  • £10.35 / Pack of 100
Estimated Shipment
23-07-2018
Add to basket
Description
Description
  • With ringmark
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Specifications
0.25%
Yes
Capillary Pipette
Glass
ISO 7550, CE, IVD 98/79 EC
Ring Mark
200μL
Videos