missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation

Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.

Brand:  Hirschmann Instruments™ 2820166

Code : A0

Additional Details : Weight : 0.03000kg

 View more versions of this product

Product Code. 10630963

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications

Specifications

DIN EN ISO 1042
Polyethylene
DURAN™
105mm
12/21 NS Standard Ground
Class A
20mL
Yes
±0.04mL
±0.04mL
Videos