Learn More
DWK Life Sciences DURAN™ Analytical Funnel, for quick filtration
The analytical funnel has a plug in its receiver which prevents the filter paper from sticking, thus enabling rapid filtration.
Brand: DWK Life Sciences 213314801
Code : A0
Additional Details : Weight : 2.21000kg
Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- For quick filtration
- The inserted glass bars make it ideal for fast filtration using paper round filters
- Fired-on, highly durable white print
Specifications
110 mm | |
Yes | |
10/Pk. |
Boro 3.3 | |
For rapid liquid filtration; The spillage of substances is avoided; Filtering |