missing translation for 'onlineSavingsMsg'
Learn More

Hamilton Tube absorption U form 125mm x 15mm length x diameter

Brand:  Hamilton ABL430025G

Tube absorption U form 125mm x 15mm length x diameter

Code : 02

Additional Details : Weight : 1.00000kg

Product Code. 12759847

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.